missing translation for 'onlineSavingsMsg'
Learn More

Baclofen, MedChemExpress

Product Code. p-200095879
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16492029

Brand: Medchem Express HYB0007

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Baclofen, a lipophilic derivative of γ-aminobutyric acid (GABA), is an orally active, selective metabotropic GABAB receptor (GABABR) agonist. Baclofen mimics the action of GABA and produces slow presynaptic inhibition through the GABAB receptor. Baclofen has high blood brain barrier penetrance. Baclofen has the potential for muscle spasticity research.

Baclofen, a lipophilic derivative of γ-aminobutyric acid (GABA), is an orally active, selective metabotropic GABAB receptor (GABABR) agonist. Baclofen mimics the action of GABA and produces slow presynaptic inhibition through the GABAB receptor. Baclofen has high blood brain barrier penetrance. Baclofen has the potential for muscle spasticity research.

Specifications

Chemical Name or Material Baclofen
Physical Form Solid
CAS 1134-47-0
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C10H12ClNO2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : 2 mg/mL (9.36 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C(O)CC(C1=CC=C(Cl)C=C1)CN
Molecular Weight (g/mol) 213.66
Formula Weight 213.66
Percent Purity 99.42%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.