missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Baclofen, MedChemExpress
Description
Baclofen, a lipophilic derivative of γ-aminobutyric acid (GABA), is an orally active, selective metabotropic GABAB receptor (GABABR) agonist. Baclofen mimics the action of GABA and produces slow presynaptic inhibition through the GABAB receptor. Baclofen has high blood brain barrier penetrance. Baclofen has the potential for muscle spasticity research.
Specifications
Specifications
| Chemical Name or Material | Baclofen |
| Physical Form | Solid |
| CAS | 1134-47-0 |
| Color | White |
| Quantity | 1 g |
| Purity Grade Notes | Research |
| Molecular Formula | C10H12ClNO2 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | H2O : 2 mg/mL (9.36 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| SMILES | O=C(O)CC(C1=CC=C(Cl)C=C1)CN |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?