missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Baclofen, Lipomed
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Baclofen
Specifications
Specifications
| Chemical Name or Material | Baclofen |
| CAS | 1134-47-0 |
| Analyte or Component Names | Baclofen |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 Cl N O2 |
| Synonym | Baclofen, (3 RS)-4-Amino-3-(4-chlorophenyl)butanoic acid |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | NCC(CC(=O)O)c1ccc(Cl)cc1 |
| IUPAC Name | 4-amino-3-(4-chlorophenyl)butanoic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction