Learn More
(+/-)-Baclofen, 99+%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J62768.03
| Quantity | 1g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 1134-47-0 | |
| MFCD00055143 | |
| UN2811 | |
| baclofen, lioresal, baclon, 4-amino-3-4-chlorophenyl butanoic acid, dl-baclofen, kemstro, gabalon, baclofene, baclofeno, baclofenum | |
| KPYSYYIEGFHWSV-UHFFFAOYSA-N | |
| 4-amino-3-(4-chlorophenyl)butanoic acid | |
| 2284 | |
| 213.66 | |
| (+/-)-Baclofen |
| C10H12ClNO2 | |
| 1g | |
| 14,937 | |
| Soluble in dilute NaOH or dilute HCl; Soluble in water at approximately 4mg/ml at pH 7;6 | |
| C1=CC(=CC=C1C(CC(=O)O)CN)Cl | |
| 213.661 | |
| CHEBI:2972 | |
| ≥99% |
Safety and Handling
- (^+)-Baclofen
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1A
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contain: Baclofen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.