missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Geobacillus stearothermophilus ATCC™ 7953™
Thermo Scientific™ Spore Strips are stabilized microorganism-impregnated strips packaged in individual recloseable plastic bags.
£941.00
Specifications
ATCC® | 7953TM* |
---|---|
Format | Spore Strips |
Product Type | Geobacillus Species |
Target Organism Class | Gram-positive Rods |
Test Type | Qualitative QC |
Product Code | Brand | ATCC® | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | ATCC® | Description | Price | Quantity & Availability | ||||
12355403
|
Thermo Scientific™
R4851001 |
7953TM* | Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 105 spore population |
£941.00
100 strips |
In Stock
Log in to see stock. |
||||
Description
Thermo Scientific™ Spore Strips are stabilized microorganism-impregnated strips packaged in individual recloseable plastic bags.
- 10 -6spore population
Specifications
7953TM* | |
Geobacillus Species | |
Qualitative QC |
Spore Strips | |
Gram-positive Rods |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.