missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Geobacillus stearothermophilus ATCC™ 7953™

Product Code. 12355403
Change view
Click to view available options
ATCC®:
7953TM*
Description:
Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 105 spore population
Unit Size:
100 strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. ATCC® Description unitSize
12355403 7953TM* Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 105 spore population 100 strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12355403 Supplier Thermo Scientific™ Supplier No. R4851001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Spore Strips are stabilized microorganism-impregnated strips packaged in individual recloseable plastic bags.

  • 10 -6spore population
TRUSTED_SUSTAINABILITY

Specifications

ATCC® 7953TM*
Description Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 105 spore population
Format Spore Strips
Product Type Quality Control Organism
Target Organism Class Gram-positive Rods
Test Type Qualitative QC

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.