missing translation for 'onlineSavingsMsg'
Learn More

3M™ Backpack Harness BPK-02

Product Code. 15461854
Change view
Click to view available options
:
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15461854 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15461854 Supplier 3M™ Supplier No. 7000044313

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

3M™ Backpack Harness BPK-02 enables the 3M™ Jupiter™ Powered Air Turbo to be carried securely on the wearers back.

  • Backpack harness enables the Powered Air Turbo to be carried on the wearer's back rather than on a belt at the waist
  • Securely holds the Powered Air Turbo on your back
  • Designed for use with the 3M™ Jupiter™ Powered Air Turbo
  • Harness with web shoulder straps and back panel for breathability and comfort
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Harness
For Use With (Equipment) Juniper™ Fan Unit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.