Learn More
Upchurch Scientific Back pressure regulator for 1/16 inch O.D. tubingSST 250psi (17bar)
Brand:  Upchurch Scientific 02017-34
Back pressure regulator for 1/16 inch O.D. tubingSST 250psi (17bar)
Product Code. 12972009
  • £443.00 / Each