missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bacitracin, zinc salt, potency min. 60 Units/mg

Product Code. 10259163 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10259163 1 g Glass Bottle 1g
10348310 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10259163 Supplier Thermo Scientific Chemicals Supplier No. 226110010
  • ONLINE EXCLUSIVE PRICE
    £26.25
    £20.65 / 1g
    Save £5.60
    21% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-89-6
MDL Number MFCD16661228
Synonym Zinc bacitracin

Specifications

Type Bacitracin Zinc Salt
Physical Form Fine Crystalline Powder
pH 6 to 7.5 (as described by Eur. Pharm.)
Melting Point 250.0°C
Infrared Spectrum Authentic
Loss on Drying 5% max. (1 g, 60°C) (P2O5) (not exceeding 0.1 kPa for 3 hrs.)
Packaging Glass Bottle
Color Beige or Gray/Yellow to White
Concentration ≥60 I.U./mg (potency, on dried subst.)
Quantity 1 g
Merck Index 15, 929
Solubility Information Solubility in water: 5.1g/L (28°C). Other solubilities: 6.55g/L methanol - 2.0g/L ethanol (28°C), 0.16g/L isopropanol- 1.3g/L ethyl acetate (28°C), 0.01g/L chloroform
Percent Purity 60 I.U./mg min. (potency, on dried subst.)
Chemical Name or Material Bacitracin, zinc salt, potency min. 60 Units/mg
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.