missing translation for 'onlineSavingsMsg'
Learn More

Bacitracin Zinc, MedChemExpress

Product Code. 16488206
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488206 100 mg 100mg
16441406 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488206 Supplier Medchem Express Supplier No. HYB0278
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bacitracin Zinc (Zinc bacitracin) is a dephosphorylation of the C55-isoprenyl pyrophosphate interference for inhibition of cleavage of Tyr from Met-enkephalin with IC50 of 10 μM.

Bacitracin Zinc (Zinc bacitracin) is a dephosphorylation of the C55-isoprenyl pyrophosphate interference for inhibition of cleavage of Tyr from Met-enkephalin with IC50 of 10 μM.

Specifications

Chemical Name or Material Bacitracin Zinc
Physical Form Solid
CAS 1405-89-6
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C66H101N17O16SZn
For Use With (Application) COVID-19-immunoregulation
Synonym Zinc bacitracin
Solubility Information 1M HCl : 50 mg/mL (33.65 mM; Need ultrasonic) ∣H2O : 1 mg/mL (0.67 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C([C@H](NC([C@@H](CCCN)NC([C@H](CCCCNC1=O)NC([C@H]([C@@H](C)CC)NC([C@@H](CCC([O-])=O)NC([C@@H](NC(C2CSC(C(C(CC)C)N)=N2)=O)CC(C)C)=O)=O)=O)=O)=O)[C@H](C)CC)N[C@@H](C(N[C@H](CC3=CN=CN3)C(N[C@H](CC([O-])=O)C(N[C@H]1CC(N)=O)=O)=O)=O)CC4=CC=CC=C4.[Zn+2]
Molecular Weight (g/mol) 1486.06
Formula Weight 1486.06
Percent Purity 98.76%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.