missing translation for 'onlineSavingsMsg'
Learn More

Bacitracin Zinc (>80%), TRC

Product Code. 17970615
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970615 50 mg 50mg
30132539 250 mg 250mg
17943425 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970615 Supplier Toronto Research Chemicals Supplier No. TRCB10650550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-89-6
Synonym Bacitracin zinc∣Baciferm (8CI)∣Albac∣Bacifarmin∣Bacifarmin-zinc∣Baciferm PB∣Noptracin∣ST 56∣Zinc-bacitracin∣Bacitracin Zinc∣GR 31385A

Specifications

Format Neat
Molecular Weight (g/mol) 1472.06
Product Type API
Quantity 50 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.