missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bacitracin, potency min. 60 Units/mg

Product Code. 10523201 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10523201

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bacitracin, CAS # 1405-87-4, is cyclic polypeptide-based antibiotic which exhibits activity against microbes causing pneumonia, skin, and eye infections.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Bacitracin is a mixture of at least nine different cyclic polypeptides, originally isolated from Bacillus bacteria in 1945

• Bacitracins work by binding divalent metal cations and preventing the hydrolysis of lipids into usable cell wall components

Applications

• Bacitracin has been paired with nanoparticles in some laboratory studies to increase its effectivity

• It is suitable for the laboratory study of bacterial resistance, infection, and cross-reactivity

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-87-4
Molecular Formula C66H103N17O16S
MDL Number MFCD00062640

Specifications

Type Bacitracin
Physical Form Powder
Melting Point 221.0°C to 225.0°C
Infrared Spectrum Authentic
Loss on Drying 5.0% max. (1 g, 60°C, 3 Hrs) (P2O5) (not exceeding 0.1 kPa)
Packaging Glass Bottle
Color Beige or White
Concentration ≥60 I.U./mg (potency, on dried subst.)
Quantity 5 g
Merck Index 15, 927
Solubility Information Solubility in water: soluble. Other solubilities: soluble in glacial acetic acid, methanol and, alcohol, practically insoluble in ether, chloroform and, acetone
Formula Weight 1422.71
Percent Purity 60 I.U./mg min. (potency, on dried subst.)
Chemical Name or Material Bacitracin
Show More Show Less
compliance-icons
Product Identifier
  • Bacitracin
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.