missing translation for 'onlineSavingsMsg'
Learn More

Bacitracin, Fisher BioReagents

Product Code. 10094627
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10094627

Brand: Fisher BioReagents BP29501

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibitor of protein disulfide isomerase. Interferes with building blocks of peptidoglycan bacterial cell wall

TRUSTED_SUSTAINABILITY

Specifications

Type Bacitracin
Physical Form Powder/Solid
CAS 1405-87-4
pH 6.0 to 7.0
Melting Point 221°C
Source Bacillus licheniformis
Loss on Drying < 5.0%
Packaging Amber Glass
Potency > 70 U/mg
Color Beige
Quantity 1 g
Molecular Formula C66H103N17O16S
Merck Index 15, 927
Solubility Information Soluble in water
Formula Weight 1422.69
Chemical Name or Material Bacitracin
Show More Show Less
compliance-icons
Product Identifier
  • Bacitracin
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.