missing translation for 'onlineSavingsMsg'
Learn More

Thermo Fisher Scientific™ Bacillus stearothermophilus ATCC® 10149™*

Product Code. 12978848
Click to view available options
Quantity:
5 Loops
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 12978848

Brand: Thermo Fisher Scientific™ R4601202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms

Recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY

Specifications

ATCC® ATCC - EULA required
Biosafety Level DG
Content And Storage 2°C to 8°C
Country of Origin US
Product Type Quality Control Organism
For Use With (Application) Ready-to-Use ATCC QC Organisms
No. per Pack 5
Packaging Type Foil Pk.
Quantity 5 Loops
Species Lactobacillus gasseri
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.