missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bacillus Spore Antigen Polyclonal Antibody, FITC

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173113
This item is not returnable.
View return policy
Description
The PA1-73113 antibody reacts with Bacillus species spores. PA1-73113 has been successfully used in Immunofluorescence applications. The PA1-73113 antibody was raised against purified spores of B. cereus (ATCC #11778) + B. subtilis (ATCC #9372).
Bacillus is a genus of rod-shaped, Gram-positive bacteria and a member of the division Firmicutes. Bacillus species are either obligate or facultative aerobes, and test positive for the enzyme catalase. Bacillus includes both free-living (nonparasitic) and parasitic pathogenic species such as B. anthracis, which causes anthrax, and B. cereus, which causes food poisoning. Under stressful environmental conditions, the bacteria can produce oval endospores that allow them to remain in a dormant state for very long periods.
Specifications
| Bacillus Spore Antigen | |
| Polyclonal | |
| FITC | |
| Anthrax Spore; Bacillus Spore | |
| Purified spores of B. cereus (ATCC #11778) + B. subtilis (ATCC #9372). | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
| Immunocytochemistry | |
| 4-5 mg/mL | |
| PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
| Rabbit | |
| IgG fraction | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction