missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bacillus cereus Selective Agar Base (Dehydrated)

Product Code. p-200112464
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10301113

Brand: Thermo Scientific™ CM0617B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selectively isolate and enumerate Bacillus cereus from food samples with this selective and differential medium.

Selectively isolate and enumerate Bacillus cereus from food samples with Thermo Scientific™ Oxoid™ Bacillus cereus Selective Agar Base (Dehydrated). Bacillus cereus Selective Agar Base meets the requirements for a medium that is sufficiently selective to be able to detect small numbers of Bacillus cereus cells and spores in the presence of large numbers of other food contaminants. The medium is also sufficiently diagnostic that colonies of Bacillus cereus are readily identified and confirmed by microscopic examination.

Bacillus cereus is a Gram-positive, rod-shaped beta hemolytic bacterium commonly found in soil and food. The role of Bacillus cereus in food poisoning, particularly from the consumption of contaminated rice, is now well documented1,2,3. The organism has also been implicated in eye infections4, 5 and a wide range of other conditions, including abscess formation, meningitis, septicemia and wound infection.

Bacillus cereus is recognized as a significant pathogen in post-operative and post-traumatic wounds of orthopedic patients6. Amongst veterinarians, Bacillus cereus is a known cause of disease, especially mastitis, in ewes and heifers7.

  • Differential: differentiates cereus from other species of Bacillus based on their resistance to polymyxin, lack of mannitol fermentation, and the presence of lecithinase.
  • Selective: inhibits the growth of gram-negative bacteria such as Escherichia coli and Pseudomonas aeruginosa due to the addition of polymixin B supplement.

Bacillus cereus Selective Agar is based on the highly specific diagnostic and selective PEMBA medium, developed by Holbrook and Anderson1 for the isolation and enumeration of Bacillus cereus in foods. It meets the requirements for a medium that is sufficiently selective to be able to detect small numbers of Bacillus cereus cells and spores in the presence of large numbers of other food contaminants. The medium is also sufficiently diagnostic that colonies of Bacillus cereus are readily identified and confirmed by microscopic examination.

The primary diagnostic features of the medium are the colonial appearance, precipitation of hydrolyzed lecithin and the failure of Bacillus cereus to utilize mannitol. The typical colonies of Bacillus cereus are crenated, about 5mm in diameter and have a distinctive turquoise to peacock blue color surrounded by a good egg yolk precipitate of the same color. These features distinguish Bacillus cereus from other Bacillus spp. except Bacillus thuringiensis.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Bacillus Cereus Selective Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 12.2L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.