missing translation for 'onlineSavingsMsg'
Learn More

Bacillol AF™ Surface Disinfectant

Product Code. 11945477
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11945477 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11945477 Supplier Bode Chemie Supplier No. 9802123
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Disinfectant with extensive spectrum of activity and alcohol based solution.

  • Ready-to-use solution
  • Rapid and comprehensive action
  • Broad material compatibility with alcohol-resistant surfaces
  • Good wetting
  • Aldehyde, colorant and fragrance-free

Specifications

Product Type Bacillol AF Disinfectant
Form Liquid
pH 6
Appearance Colorless
Certifications/Compliance CE, MDD, VAH
Composition Active ingredients: Propan-1-ol 400mg/g, propan-2-ol 250mg/g, Ethanol 47mg/g
Container Type Bottle
For Use With (Application) For areas that require quick exposure and drying times and residue free drying
Volume (Metric) 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.