missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACH2 Antibody (PCRP-BACH2-5B11), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45071
This item is not returnable.
View return policy
Description
BACH2 Monoclonal antibody specifically detects BACH2 in Human samples. It is validated for Flow Cytometry
Specifications
| BACH2 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry : 1-2ug/million cells | |
| BTB and CNC homolog 2, BTB and CNC homology 1, basic leucine zipper transcription factor 2, transcription regulator protein BACH2 | |
| Recombinant fragment (around aa11-132) of human BACH2 protein (Uniprot: Q9BYV9). Exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry | |
| PCRP-BACH2-5B11 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 60468 | |
| Store at -20 to -84°C. Avoid freeze-thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur