missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACH1 Antibody (OTI4E11), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70240PECY55
This item is not returnable.
View return policy
Description
BACH1 Monoclonal specifically detects BACH1 in Human samples. It is validated for Flow Cytometry.
Specifications
| BACH1 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| BACH1 | |
| Full length recombinant protein of human BACH1 (NP_996749) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI4E11 | |
| Flow Cytometry | |
| basic region leucine zipper transcriptional regulator BACH1, BTB and CNC homolog 1, BTB and CNC homology 1, basic leucine zipper transcription factor 1, HA2303, transcription regulator protein BACH1, transcription regulator protein10BACH-1 | |
| Mouse | |
| Affinity Purified | |
| Breast Cancer, Cancer, DNA Repair | |
| 571.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction