missing translation for 'onlineSavingsMsg'
Learn More

BACH1 Antibody (OTI4E11), DyLight 594, Novus Biologicals™

Product Code. 18428112 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428112 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428112 Supplier Novus Biologicals Supplier No. NBP270240DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BACH1 Monoclonal antibody specifically detects BACH1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen BACH1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI4E11
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias basic region leucine zipper transcriptional regulator BACH1, BTB and CNC homolog 1, BTB and CNC homology 1, basic leucine zipper transcription factor 1, HA2303, transcription regulator protein BACH1, transcription regulator protein10BACH-1
Host Species Mouse
Immunogen Full length recombinant protein of human BACH1 (NP_996749) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 571
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.