missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACE1 Rabbit anti-Human, Mouse, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12807-1-AP-20UL
This item is not returnable.
View return policy
Description
Accumulation of the amyloid-b (Ab) plaque in the cerebral cortex is a critical event in the pathogenesis Alzheimer′s disease. Ab peptide is generated by proteolytic cleavage of the b-amyloid protein precursor(APP) at b- and g-sites by two proteases. APP is first cleaved by b-secretase, producing a soluble derivative of the protein and a membrane anchored 99-amino acid carboxy-terminal fragment (C99). The C99 fragment serves as substrate for g-secretase to generate the 4 kDa amyloid-b peptide, which is deposited in the brains of all sufferers of Alzheimer′s disease. The long-sought b-secretase was recently identified by several groups independently and designated beta-site APP cleaving enzyme (BACE) and aspartyl protease 2 (Asp2). bACE/Asp2 is a novel transmembrane aspartic protease and colocalizes with APP.Specifications
| BACE1 | |
| Polyclonal | |
| Unconjugated | |
| BACE1 | |
| Asp 2, ASP2, Aspartyl protease 2, BACE, BACE1, Beta secretase 1, HSPC104, KIAA1149, Memapsin 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23621, 23821, 403005 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P56817, P56818 | |
| bace1 | |
| BACE1 Fusion Protein Ag3559 | |
| 20 μL | |
| Primary | |
| Mouse, Zebrafish, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction