missing translation for 'onlineSavingsMsg'
Learn More

BACE-2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18407083 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407083 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407083 Supplier Novus Biologicals Supplier No. NBP177309MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BACE-2 Polyclonal antibody specifically detects BACE-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BACE-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AEPLC, Asp 1, ASP1, ASP21, Aspartic-like protease 56 kDa, Aspartyl protease 1, beta secretase 2, beta-secretase 2, beta-site amyloid beta A4 precursor protein-cleaving enzyme 2, Beta-site amyloid precursor protein cleaving enzyme 2, Beta-site APP cleaving enzyme 2, beta-site APP-cleaving enzyme 2, beta-site APP-cleaving enzyme 2, EC 3.4.2310ALP5656 kDa aspartic-like protease, CEAP1, Down region aspartic protease, Down syndrome region aspartic protease, DRAPBAE2, EC 3.4.23, EC 3.4.23.45, Memapsin-1, Membrane-associated aspartic protease 1, Theta-secretase, transmembrane aspartic proteinase Asp1
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the amino terminus of human BACE2. The immunogen is located within amino acids 30 - 80 of BACE2. Amino Acid Squence: APTPGPGTPAERHADG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 25825
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.