missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACE-1 Antibody (S03-9C8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19577-50ul
This item is not returnable.
View return policy
Description
BACE-1 Monoclonal antibody specifically detects BACE-1 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| BACE-1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| S03-9C8 | |
| Unconjugated | |
| APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2 | |
| Recombinant protein of mouse BACE-1 (Uniprot # P56818) | |
| 50 μg | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 23621 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction