missing translation for 'onlineSavingsMsg'
Learn More

BACE-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-77307-0.025mg

 View more versions of this product

Product Code. 18732954

  • £141.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

BACE-1 Polyclonal specifically detects BACE-1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BACE-1
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2
Rabbit
55 kDa
0.025 mg
Neurodegeneration, Neuroscience
23621
Human, Mouse
IgG
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 10-20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
AF190725
BACE1
Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human BACE. The immunogen is located within the last 50 amino acids of BACE. Amino Acid Squence: CLRQQHDDFADDISLL
Ion exchange chromatography
RUO
Primary
Human BACE-1 has 6 isoforms, including isoform A (501aa, 55.8kD), isoform B (476aa, 52.9kD), isoform C (457aa, 51.1kD), isoform D (432aa, 48.2kD), isoform 5 (401aa, 45kD) and isoform 6 (376aa, 42.2kD). This antibody detects all human isoforms. Mouse BACE-1 has only 1 isoform (501aa, 55.7kD). Rat BACE-1 has only one isoform identified so far (501aa, 55.8kD).
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only