missing translation for 'onlineSavingsMsg'
Learn More

BACE-1 Antibody (027), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18334870 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334870 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334870 Supplier Novus Biologicals Supplier No. NBP289254AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BACE-1 Monoclonal specifically detects BACE-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BACE-1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 27
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human BACE-1 (NP_036236.1; Met1-Thr457).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23621
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.