missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bac-to-Bac™ HBM TOPO™ Cloning Kit

Product Code. 10758415
Change view
Click to view available options
No. of Reactions:
20
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10758415 20 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10758415 Supplier Invitrogen™ Supplier No. A11338

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Bac-to-Bac™ HBM TOPO™ Cloning Kit

Bac-to-Bac™ Baculovirus Expression is an efficient method for producing baculovirus for high-level protein expression in insect cells. It relies on generation of recombinant virus by site-specific transposition in E. coli (DH10Bac™) rather than homologous recombination in insect cells. The new pFastBac™ HBM TOPO™ vector enables secreted protein expression because it has the honeybee melittin (HBM) secretion signal. The new vector should be strongly considered for the expression of glycoproteins. Glycoproteins cannot be glycosylated in the absence of any secretion signal. In contrast to glycoproteins secreted from mammalian cells, glycoproteins secreted from baculovirus can be easily de-glycosylated in vitro. This is a very important feature in order to crystallize proteins!

The pFastBac™ HBM TOPO™ vector has also a C-terminal His-Tag with a TEV cleavage signal to enable easy purification of Histidine fusion proteins on nickel-chelating resins (ProBond™ Purification System) and native proteins with the aid of AcTev™ protease.
The vector uses the strong polyhedrin promoter to generate high levels of expression in a variety of insect cell lines such as Sf9, Mimic™ Sf9, Sf21, and High Five™ cells in Sf-900 II & Sf-900™ III media from Gibco.

Traditional baculovirus systems require purification and amplification of an initial low-titer viral supernatant. This requires a time-consuming plaque assay. Bac-to-Bac™'s pFastBac™ vectors, however, recombines with the parent bacmid in DH10Bac™ E. coli competent cells to form an expression bacmid. Transfect the bacmid into insect cells for fast production of a high titer of pure recombinant baculovirus particles in the very first transfection. You'll save weeks of precious time. Collect a pure P2 baculovirus stock on Day 10 without the necessity of tedious plaques assay.
TRUSTED_SUSTAINABILITY

Specifications

Cell Line High Five™, Sf21, Sf9
Cell Type Insect Cell
Cloning Method Blunt TOPO
Cleavage TEV Protease Recognition Site
Content And Storage Bac-to-Bac™ HBM TOPO™ Cloning Kit contains: (For details see the manual)
• Vector kit for 20 topo cloning reactions
• pFastBac⁄HBM TOPO™ Vector containing the C-terminal TEV cleavage sit and His-Tag.
• pFastBac⁄CT-Gus Control Plasmid (Control expression vector)
• Other reagents supplied: 10x PCR, dNTP Mix, Salt Solution, Sterile Water, Control PCR template, Polyhedrin Forward Primer, SV40 pA Reverse Primer
• Kit containing competent cells (20 reactions):
• One Shot™ Mach1-T1R Chemically Competent E. coli

Vector kit: store at -20°C
One Shot™ Mach1-T1R Chemically Competent E. coli: store at -80°C
Format Kit
Expression Mechanism Cell-Based Expression
Expression System Baculovirus
For Use With (Application) Protein Expression
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual)
Key Functions Secreted Expression
No. of Reactions 20
Product Line Bac-to-Bac, TOPO
Product Type Cloning Kit
Promoter Polyhedrin
Protein Tag HBM (Honeybee Melittin), His Tag (6x)
Selection Agent (Eukaryotic) None
Vector pFastBac/HBM-TOPO
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.