missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Baboon Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-665
This item is not returnable.
View return policy
Description
Baboon Polyclonal specifically detects Baboon in Drosophila samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Baboon | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Drosophila | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.75 mg/mL | |
| Western Blot 1:200, Immunoprecipitation 1:200 | |
| Activin A receptor 45A, ATR 1, Atr 45A, Atr I, ATR1, Atr45A, AtrI, Bab, Babo, CG8224, G protein coupled receptor kinase 3, Gprk 3, Gprk3, l(2)k16912, RE55648p, STK E, STKE | |
| Synthetic peptide: QPDPSIEEMKKVVCIEKCRPN, corresponding to amino acids 552 to 572 of Baboon.The was generated against a peptide sequence near the C terminal end of the protein.The peptides was amidated for conjugation. | |
| 0.1 mg | |
| Signal Transduction | |
| The is unique to Baboon protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction