missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAAT1 Polyclonal Antibody

BAAT1 Polyclonal Antibody
Brand: Invitrogen PA598531
This item is not returnable.
View return policy
Description
BAAT1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| BAAT1 | |
| Polyclonal | |
| Unconjugated | |
| BRAT1 | |
| AA881470; BAAT1; Brat1; Brat1-ps1; BRCA1 associated ATM activator 1; BRCA1-associated ATM activator 1; BRCA1-associated ATM activator 1, pseudogene 1; BRCA1-associated protein required for ATM activation protein 1; C7orf27; HEAT repeat-containing protein C7orf27; MGC22916; RGD1565146; RMFSL | |
| Rabbit | |
| Protein G | |
| RUO | |
| 221927 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.95 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6PJG6 | |
| BRAT1 | |
| Recombinant Human BRCA1-associated ATM activator 1 protein (603-821AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction