missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAAT1 Antibody (JE64-71), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32089
This item is not returnable.
View return policy
Description
BAAT1 Monoclonal antibody specifically detects BAAT1 in Human samples. It is validated for Western Blot
Specifications
| BAAT1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 | |
| BAAT1, BRCA1-associated ATM activator 1, BRCA1-associated protein required for ATM activation protein 1, C7orf27, chromosome 7 open reading frame 27, HEAT repeat-containing protein C7orf27, MGC22916 | |
| Recombinant protein within human BAAT1 aa 672-821/821. (Uniprot: Q6PJG6) | |
| 100 μL | |
| Breast Cancer, Cancer, DNA Repair | |
| 221927 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE64-71 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction