missing translation for 'onlineSavingsMsg'
Learn More

BA Antigen (Leu21) Antibody (DF1588), DyLight 350, Novus Biologicals™

Product Code. 18400615 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400615 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400615 Supplier Novus Biologicals Supplier No. NBP254564UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BA Antigen (Leu21) Monoclonal antibody specifically detects BA Antigen (Leu21) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen BA Antigen (Leu21)
Applications Flow Cytometry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone DF1588
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias CD78, CD78, Cdw78, Cdw78, LO panB a
Host Species Mouse
Immunogen Human leucocytes
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.