missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B9D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599068
This item is not returnable.
View return policy
Description
B9D1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a B9 domain-containing protein, one of several that are involved in ciliogenesis. Alterations in expression of this gene have been found in a family with Meckel syndrome. Meckel syndrome has been associated with at least six different genes. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Three alternatively spliced transcript variants that encode different proteins have been described for this gene.
Specifications
| B9D1 | |
| Polyclonal | |
| Unconjugated | |
| B9D1 | |
| AW045994; B9; B9 domain containing 1; B9 domain-containing protein 1; B9 protein domain 1; B9D1; Endothelial precursor cells protein B9; endothelial precursor protein B9; Eppb9; MKS1-related protein 1; MKS9; MKSR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27077, 27078 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9R1S0, Q9UPM9 | |
| B9D1 | |
| Recombinant Human B9 domain-containing protein 1 protein (1-130AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction