missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B9D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572758
This item is not returnable.
View return policy
Description
B9D1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
This gene encodes a B9 domain-containing protein, one of several that are involved in ciliogenesis. Alterations in expression of this gene have been found in a family with Meckel syndrome. Meckel syndrome has been associated with at least six different genes. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Three alternatively spliced transcript variants that encode different proteins have been described for this gene.
Specifications
| B9D1 | |
| Polyclonal | |
| Unconjugated | |
| B9D1 | |
| AW045994; B9; B9 domain containing 1; B9 domain-containing protein 1; B9 protein domain 1; B9D1; Endothelial precursor cells protein B9; endothelial precursor protein B9; Eppb9; MKS1-related protein 1; MKS9; MKSR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27077 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9UPM9 | |
| B9D1 | |
| B9D1 antibody for an 18 amino acid synthetic peptide near the carboxy terminus of human B9D1. The immunogen is within amino acids 130 - 180 of B9D1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction