missing translation for 'onlineSavingsMsg'
Learn More

B9D1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18405568 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405568 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405568 Supplier Novus Biologicals Supplier No. NBP281899MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

B9D1 Polyclonal antibody specifically detects B9D1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen B9D1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias B9, B9 domain-containing protein 1, B9 protein domain 1, endothelial precursor protein B9, EPPB9, MKS1-related protein 1, MKSR1
Host Species Rabbit
Immunogen B9D1 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human B9D1. The immunogen is located within amino acids 130 - 180 of B9D1. Amino Acid Squence: RRPEYTDPKVVAQGEGRE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 27077
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.