missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H7/HHLA2 Antibody (907834), Alexa Fluor™ 532, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB80842AF532
This item is not returnable.
View return policy
Description
B7-H7/HHLA2 Monoclonal specifically detects B7-H7/HHLA2 in Human samples. It is validated for Flow Cytometry.
Specifications
| B7-H7/HHLA2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HHLA2 | |
| Partial recombinant human B7-H7 protein (amino acids 1-344) [UniProt Q9UM44] | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 907834 | |
| Flow Cytometry | |
| HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
| Mouse | |
| 0.1 mL | |
| 11148 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction