missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H6 Mouse anti-Human, Clone: B7H6/4821, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07759-100ug
This item is not returnable.
View return policy
Description
B7-H6 Monoclonal specifically detects B7-H6 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| B7-H6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| B7 homolog 6, B7-H6, DKFZp686O24166 | |
| Recombinant fragment (around aa38-169) of human B7-H6 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Immunology | |
| 374383 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| B7H6/4821 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction