missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H6 Antibody (Dartmouth patent anti-B7-H6), DyLight 550, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27971R
This item is not returnable.
View return policy
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| B7-H6 | |
| Monoclonal | |
| DyLight 550 | |
| B7 homolog 6, B7-H6, DKFZp686O24166 | |
| B7-H6/ NCR3LG1 | |
| 0.1 mL | |
| Cancer, Immunology | |
| 374383 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, ELISA, Functional Assay | |
| Dartmouth patent anti-B7-H6 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction