missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ B7-H4 Antibody (Millennium patent anti-B7-H4), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28513-50ug
This item is not returnable.
View return policy
Description
B7-H4 Monoclonal antibody specifically detects B7-H4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| B7-H4 | |
| Monoclonal | |
| Unconjugated | |
| Q7Z7D3 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Millennium patent anti-B7-H4 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| B7h.5, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1VCTN1, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1 | |
| B7-H4/ VTCN1 | |
| 50 μg | |
| Cancer, Cell Cycle and Replication | |
| 79679 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction