missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
B7-H3/CD276 Monoclonal antibody specifically detects B7-H3/CD276 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | B7-H3/CD276 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | BLR025F |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 3 μL/mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500 |
| Formulation | Borate Buffered Saline (BBS) pH 8.2 with, BSA Free with 0.09% Sodium Azide |
| Gene Alias | B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule |
| Host Species | Rabbit |
| Immunogen | Immunogen was a peptide representing a region between residue 500 and the C-terminus (residue 534) of human CD276 Molecule using the numbering given in entry NP_001019907.1 (GeneID 80381). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?