missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
B7-H3/CD276 Polyclonal antibody specifically detects B7-H3/CD276 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | B7-H3/CD276 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 350 to 400 of human CD276 Molecule using the numbering given in entry NP_001019907.1 (GeneID 80381). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?