missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£244.00 - £534.00
Specifications
| Antigen | PD-L1/B7-H1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 0.5 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1-5 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount, Immunofluorescence 20 μg/mL, Knockdown Validated, Dual RNAscope ISH-IHC |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown, In Situ Hybridization (ISH) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18749363
|
Novus Biologicals
NBP1-76769SS |
0.025 mg |
£244.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18725593
|
Novus Biologicals
NBP1-76769 |
0.1 mg |
£534.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PD-L1 Polyclonal specifically detects PD-L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence, Knockdown Validated, Dual RNAscope ISH-IHC.Specifications
| PD-L1/B7-H1 | |
| Western Blot 1:1000, Flow Cytometry 0.5 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1-5 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount, Immunofluorescence 20 μg/mL, Knockdown Validated, Dual RNAscope ISH-IHC | |
| Polyclonal | |
| Rabbit | |
| Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| CD274 | |
| IgG | |
| Affinity Purified | |
| PD-L1 antibody has no cross-reactivity to PD-L2. |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown, In Situ Hybridization (ISH) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| NP_054862 | |
| 29126 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the center of human PD-L1. The immunogen is located within amino acids 60-110 of PD-L1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 37 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title