missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-1/CD80, Human anti-Human, Clone: IDEC-114 (Galiximab), Novus Biologicals™
Shop All Bio Techne ProductsDescription
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Functional, Block/Neutralize.
Specifications
Specifications
| Antigen | B7-1/CD80 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Blocking Assay |
| Classification | Monoclonal |
| Clone | IDEC-114 (Galiximab) |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Block/Neutralize |
| Formulation | PBS |
| Gene Alias | Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 |
| Gene Symbols | CD80 |
| Show More |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?