missing translation for 'onlineSavingsMsg'
Learn More

B7-1/CD80 Human anti-Human, Clone: 16C10, Novus Biologicals™

Product Code. 18372573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372573 0.025 mg 25µg
18336023 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18372573 Supplier Novus Biologicals Supplier No. NBP3120260.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human samples. It is validated for ELISA, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen B7-1/CD80
Applications ELISA, Neutralization
Classification Monoclonal
Clone 16C10
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA, Block/Neutralize
Formulation PBS
Gene Alias Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Host Species Human
Immunogen This antibody was generated by immunization of cynomolgus monkeys with human B7.1 antigen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 941
Target Species Human
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.