missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | B7-1/CD80 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | RM80 |
| Conjugate | PE |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 |
| Host Species | Rat |
| Immunogen | This antibody was raised by immunising Sprague Dawley rats with a mouse B cell line, BCL1 and fusing immune splenocytes with P3U1 myeloma cells. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?