missing translation for 'onlineSavingsMsg'
Learn More

B7-1/CD80 Antibody (IDEC-114 (Galiximab)), Biotin, Novus Biologicals™

Product Code. 18344550 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344550 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344550 Supplier Novus Biologicals Supplier No. NBP281100B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B7-1/CD80
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone IDEC-114 (Galiximab)
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Gene Symbols CD80
Host Species Rabbit
Immunogen Galiximab was prepared by immunizing cynomolgus monkeys with recombinant B7-1/CD80 antigen. The variable regions of the light and heavy chains were then cloned by being incorporated into a cassette vector (N5LG1) containing human constant region genes and subsequently transfected into the Dg44 CHO cell line (Hariharan K, 2013)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 941
Test Specificity Galiximab is a primatized mAb which consists of human constant and primate (cynomolgus macaque) variable regions and binds specifically to B7-1/CD80. B7-1/CD80 is a surface glycoprotein and a member of the B7 family of costimulatory molecules. B7-1/CD80 antigen regulates T cell activation (through interacting with CD28 or CD152) and is expressed transiently in antigen-presenting cells, T cells and normal B cells, and expressed consitutively on various subtypes of B-cell lymphomas.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.