missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human samples. It is validated for ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | B7-1/CD80 |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 16C10 |
| Conjugate | Alexa Fluor 750 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 |
| Host Species | Rabbit |
| Immunogen | This antibody was generated by immunization of cynomolgus monkeys with human B7.1 antigen. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?