missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B4GALT7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103657
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total B4GALT7.
Required for the biosynthesis of the tetrasaccharide linkage region of proteoglycans, especially for small proteoglycans in skin fibroblasts.
Specifications
| B4GALT7 | |
| Polyclonal | |
| Unconjugated | |
| B4GALT7 | |
| B4galt7; b4Gal-T7; Beta-1,4-galactosyltransferase 7; beta-1,4-galactosyltransferase VII; beta-1,4-GalTase 7; beta4Gal-T7; beta4GalT-VII; EDSP1; EDSSLA; galactosyltransferase I; proteoglycan UDP-galactose:beta-xylose beta1,4-galactosyltransferase I; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 7; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 7; UDP-galactose:beta-xylose beta-1,4-galactosyltransferase; UNQ748/PRO1478; XGALT1; XGalT-1; XGPT; XGPT1; Xylosylprotein 4-beta-galactosyltransferase; xylosylprotein beta 1,4-galactosyltransferase, polypeptide 7; Xylosylprotein beta-1,4-galactosyltransferase; xylosylprotein beta1,4-galactosyltransferase, polypeptide 7 (galactosyltransferase I) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11285, 218271, 364675 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R087, Q9UBV7 | |
| B4GALT7 | |
| A synthesized peptide derived from human B4GALT7(Accession Q9UBV7), corresponding to amino acid residues I203-K253. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction