missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ B4GALT7 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA559949

Product Code. 15797791

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PEHWEEDASW GPHRLAVLVP FRERFEELLV FVPHMRRFLS RKKIRHHIYV LNQVDHFRFN RAALINVGFL ESSNSTDYIA MHDVDLLPLN EELDYGFPEA GPFHVASPEL HPLYHYKTYV G Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

Required for the biosynthesis of the tetrasaccharide linkage region of proteoglycans, especially for small proteoglycans in skin fibroblasts.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

B4GALT7
Polyclonal
Unconjugated
B4GALT7
B4galt7; b4Gal-T7; Beta-1,4-galactosyltransferase 7; beta-1,4-galactosyltransferase VII; beta-1,4-GalTase 7; beta4Gal-T7; beta4GalT-VII; EDSP1; EDSSLA; galactosyltransferase I; proteoglycan UDP-galactose:beta-xylose beta1,4-galactosyltransferase I; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 7; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 7; UDP-galactose:beta-xylose beta-1,4-galactosyltransferase; UNQ748/PRO1478; XGALT1; XGalT-1; XGPT; XGPT1; Xylosylprotein 4-beta-galactosyltransferase; xylosylprotein beta 1,4-galactosyltransferase, polypeptide 7; Xylosylprotein beta-1,4-galactosyltransferase; xylosylprotein beta1,4-galactosyltransferase, polypeptide 7 (galactosyltransferase I)
Rabbit
Antigen affinity chromatography
RUO
11285
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9UBV7
B4GALT7
Recombinant protein corresponding to Human B4GALT7. Recombinant protein control fragment (Product #RP-89958).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.