missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B4GALT6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99252-100ul
This item is not returnable.
View return policy
Description
B4GALT6 Polyclonal antibody specifically detects B4GALT6 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| B4GALT6 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| B4Gal-T6, beta-1,4-galactosyltransferase 6, Beta-1,4-GalTase 6, Beta4Gal-T6, beta4GalT-VI, EC 2.4.1.-, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 6, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 6, UDP-Gal:glucosylceramide beta-1,4-galactosyltransferase, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 6 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse B4GALT6. | |
| 100 μg | |
| Lipid and Metabolism | |
| 9331 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction