Learn More
B4GALT4 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30478-1-AP-150UL
Description
The B4GALT4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human B4GALT4. This antibody recognizes human, mouse, rat antigen. The B4GALT4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| B4GALT4 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| B4Gal T4, B4GALT4, Beta 1,4 GalTase 4, beta4Gal T4 | |
| 150 μL | |
| 8702 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |