missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B4GALT4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£140.00 - £283.00
Specifications
| Antigen | B4GALT4 |
|---|---|
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18321745
|
Novus Biologicals
NBP3-15515-20UL |
20 μg |
£140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18333643
|
Novus Biologicals
NBP3-15515-100UL |
100 μg |
£283.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B4GALT4 Polyclonal antibody specifically detects B4GALT4 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| B4GALT4 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| B4Gal-T4, beta-1,4-galactosyltransferase 4, Beta-1,4-GalTase 4, beta4Gal-T4, beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 4, EC 2.4.1.-, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 4, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 4, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 4, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 35-344 of human B4GALT4 (NP_003769.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| 8702 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title